siRNA Details
| Gene Symbol: Olfr173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147000.2 | NP_667211.2 | 1 | 854 | ||
| GenBank mRNA | BC120572.1 | - | 1 | 854 | 40129495 | |
| BC137837.1 | - | 1 | 854 | 8860856 | ||
| Gene Symbol: Olfr172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147001.2 | NP_667212.2 | 1 | 822 | ||
| GenBank mRNA | BC131922.1 | - | 1 | 822 | 40130199 | |
| BC131948.1 | - | 1 | 821 | 40130225 | ||

Olfr173
Genomic Map