siRNA Details
| Gene Symbol: Olfr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147003.1 | NP_667214.1 | 1 | 759 | ||
| GenBank mRNA | BC104101.2 | - | 1 | 759 | 40043110 | |
| BC104102.1 | - | 1 | 759 | 40043111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr139| Gene Symbol: Olfr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147003.1 | NP_667214.1 | 1 | 759 | ||
| GenBank mRNA | BC104101.2 | - | 1 | 759 | 40043110 | |
| BC104102.1 | - | 1 | 759 | 40043111 | ||