siRNA Details
| Gene Symbol: Olfr395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147005.1 | NP_667216.1 | 1 | 241 | ||
| Gene Symbol: Olfr392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147006.2 | NP_667217.2 | 2 | 371 | ||
| GenBank mRNA | CB173600.1 | - | 1 | 62 | ||
| DR065907.1 | - | 2 | 374 | |||

Olfr395
Genomic Map