siRNA Details
| Gene Symbol: Olfr392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147006.2 | NP_667217.2 | 2 | 896 | ||
| GenBank mRNA | CB173273.1 | - | 1 | 344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr392| Gene Symbol: Olfr392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147006.2 | NP_667217.2 | 2 | 896 | ||
| GenBank mRNA | CB173273.1 | - | 1 | 344 | ||