siRNA Details
Gene Symbol: Olfr392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147006.2 | NP_667217.2 | 2 | 767 | ||
GenBank mRNA | CB173273.1 | - | 1 | 215 | ||
CB173600.1 | - | 1 | 458 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147006.2 | NP_667217.2 | 2 | 767 | ||
GenBank mRNA | CB173273.1 | - | 1 | 215 | ||
CB173600.1 | - | 1 | 458 |