siRNA Details
Gene Symbol: Olfr1052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147010.2 | NP_667221.2 | 1 | 157 | ||
GenBank mRNA | BC127979.1 | - | 1 | 161 | 40135213 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147010.2 | NP_667221.2 | 1 | 157 | ||
GenBank mRNA | BC127979.1 | - | 1 | 161 | 40135213 |