siRNA Details
| Gene Symbol: Olfr1044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147011.1 | NP_667222.1 | 1 | 902 | ||
| GenBank mRNA | BC111826.1 | - | 1 | 901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1044| Gene Symbol: Olfr1044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147011.1 | NP_667222.1 | 1 | 902 | ||
| GenBank mRNA | BC111826.1 | - | 1 | 901 | ||