siRNA Details
Gene Symbol: Olfr1038-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147013.2 | NP_667224.1 | 2 | 740 | ||
GenBank mRNA | CB173938.1 | - | 1 | 52 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1038-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147013.2 | NP_667224.1 | 2 | 740 | ||
GenBank mRNA | CB173938.1 | - | 1 | 52 |