siRNA Details
| Gene Symbol: Olfr1019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147015.1 | NP_667226.1 | 1 | 115 | ||
| GenBank mRNA | BC117705.1 | - | 1 | 115 | 40081144 | |
| BC117706.1 | - | 1 | 115 | 40081145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1019| Gene Symbol: Olfr1019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147015.1 | NP_667226.1 | 1 | 115 | ||
| GenBank mRNA | BC117705.1 | - | 1 | 115 | 40081144 | |
| BC117706.1 | - | 1 | 115 | 40081145 | ||