siRNA Details
| Gene Symbol: Olfr1056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147018.2 | NP_667229.2 | 1 | 386 | ||
| GenBank mRNA | BC145903.1 | - | 1 | 386 | 40131206 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1056| Gene Symbol: Olfr1056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147018.2 | NP_667229.2 | 1 | 386 | ||
| GenBank mRNA | BC145903.1 | - | 1 | 386 | 40131206 | |