siRNA Details
| Gene Symbol: Olfr378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147024.2 | NP_667235.1 | 2 | 1019 | ||
| GenBank mRNA | BC139813.1 | - | 1 | 888 | 40135620 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr378| Gene Symbol: Olfr378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147024.2 | NP_667235.1 | 2 | 1019 | ||
| GenBank mRNA | BC139813.1 | - | 1 | 888 | 40135620 | |