siRNA Details
| Gene Symbol: Olfr1120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147029.1 | NP_667240.1 | 1 | 264 | ||
| GenBank mRNA | BC119117.1 | - | 1 | 284 | 8733866 | |
| BC119143.1 | - | 1 | 284 | 8733892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1120| Gene Symbol: Olfr1120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147029.1 | NP_667240.1 | 1 | 264 | ||
| GenBank mRNA | BC119117.1 | - | 1 | 284 | 8733866 | |
| BC119143.1 | - | 1 | 284 | 8733892 | ||