siRNA Details
| Gene Symbol: Olfr1134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC125403.1 | - | 1 | 429 | 40129918 | |
| BC125405.1 | - | 1 | 429 | 40129920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1134| Gene Symbol: Olfr1134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC125403.1 | - | 1 | 429 | 40129918 | |
| BC125405.1 | - | 1 | 429 | 40129920 | ||