siRNA Details
| Gene Symbol: Olfr705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147032.2 | NP_667243.2 | 1 | 500 | ||
| GenBank mRNA | BC111577.1 | - | 1 | 499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr705| Gene Symbol: Olfr705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147032.2 | NP_667243.2 | 1 | 500 | ||
| GenBank mRNA | BC111577.1 | - | 1 | 499 | ||