siRNA Details
| Gene Symbol: Olfr713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147034.1 | NP_667245.1 | 1 | 48 | ||
| GenBank mRNA | BC100704.1 | - | 1 | 47 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr713| Gene Symbol: Olfr713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147034.1 | NP_667245.1 | 1 | 48 | ||
| GenBank mRNA | BC100704.1 | - | 1 | 47 | ||