siRNA Details
Gene Symbol: Olfr679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147044.1 | NP_667255.1 | 1 | 606 | ||
GenBank mRNA | BC127993.1 | - | 1 | 610 | 40135720 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147044.1 | NP_667255.1 | 1 | 606 | ||
GenBank mRNA | BC127993.1 | - | 1 | 610 | 40135720 |