siRNA Details
Gene Symbol: Olfr683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147045.1 | NP_667256.1 | 1 | 487 | ||
GenBank mRNA | AF121981.1 | - | 1 | 80 | ||
BC111830.1 | - | 1 | 480 | |||
BC148203.1 | - | 1 | 487 | 40055104 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
