siRNA Details
| Gene Symbol: Olfr659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147050.1 | NP_667261.1 | 1 | 235 | ||
| GenBank mRNA | BC148221.1 | - | 1 | 239 | 40136035 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr659| Gene Symbol: Olfr659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147050.1 | NP_667261.1 | 1 | 235 | ||
| GenBank mRNA | BC148221.1 | - | 1 | 239 | 40136035 | |