siRNA Details
Gene Symbol: Olfr362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147051.1 | NP_667262.1 | 1 | 688 | ||
GenBank mRNA | BC104107.1 | - | 1 | 688 | 40043427 | |
BC104108.1 | - | 1 | 688 | 40043429 | ||
BC106827.1 | - | 1 | 688 | 40044131 |
