siRNA Details
| Gene Symbol: Olfr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147053.1 | NP_667264.1 | 1 | 98 | ||
| GenBank mRNA | BC107338.1 | - | 1 | 98 | ||
| BC107339.1 | - | 1 | 98 | |||
| Gene Symbol: Olfr584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147054.1 | NP_667265.1 | 1 | 98 | ||
| GenBank mRNA | BC128007.1 | - | 1 | 102 | 40135987 | |

Olfr582
Genomic Map