siRNA Details
| Gene Symbol: Olfr691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147061.1 | NP_667272.1 | 1 | 927 | ||
| GenBank mRNA | BC104116.2 | - | 1 | 919 | ||
| BC104117.1 | - | 1 | 919 | |||
| BC119417.2 | - | 1 | 927 | 40044083 | ||
| BC119418.1 | - | 1 | 927 | 40044084 | ||

Olfr691
Genomic Map