siRNA Details
| Gene Symbol: Olfr124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147062.2 | NP_667273.1 | 1 | 756 | ||
| GenBank mRNA | BC119256.1 | - | 1 | 756 | 8734005 | |
| BC119258.1 | - | 1 | 756 | 8734007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr124| Gene Symbol: Olfr124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147062.2 | NP_667273.1 | 1 | 756 | ||
| GenBank mRNA | BC119256.1 | - | 1 | 756 | 8734005 | |
| BC119258.1 | - | 1 | 756 | 8734007 | ||