siRNA Details
Gene Symbol: Olfr124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147062.2 | NP_667273.1 | 1 | 850 | ||
GenBank mRNA | BC119256.1 | - | 1 | 850 | 8734005 | |
BC119258.1 | - | 1 | 850 | 8734007 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147062.2 | NP_667273.1 | 1 | 850 | ||
GenBank mRNA | BC119256.1 | - | 1 | 850 | 8734005 | |
BC119258.1 | - | 1 | 850 | 8734007 |