siRNA Details
| Gene Symbol: Olfr1389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147066.2 | NP_667277.1 | 1 | 814 | ||
| GenBank mRNA | BC120728.1 | - | 1 | 814 | 40129651 | |
| BC120730.1 | - | 1 | 814 | 40129653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1389| Gene Symbol: Olfr1389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147066.2 | NP_667277.1 | 1 | 814 | ||
| GenBank mRNA | BC120728.1 | - | 1 | 814 | 40129651 | |
| BC120730.1 | - | 1 | 814 | 40129653 | ||