siRNA Details
| Gene Symbol: Olfr1062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147078.2 | NP_667289.2 | 1 | 651 | ||
| Gene Symbol: Olfr1057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207563.2 | NP_997446.1 | 1 | 651 | ||

Olfr1062
Genomic Map