siRNA Details
| Gene Symbol: Olfr609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147082.2 | NP_667293.2 | 1 | 244 | ||
| GenBank mRNA | BC128008.1 | - | 1 | 248 | 40135990 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr609| Gene Symbol: Olfr609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147082.2 | NP_667293.2 | 1 | 244 | ||
| GenBank mRNA | BC128008.1 | - | 1 | 248 | 40135990 | |