siRNA Details
| Gene Symbol: Olfr622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147083.1 | NP_667294.1 | 1 | 701 | ||
| GenBank mRNA | BC132174.1 | - | 1 | 799 | 40130451 | |
| BC132176.1 | - | 1 | 799 | 40130453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr622| Gene Symbol: Olfr622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147083.1 | NP_667294.1 | 1 | 701 | ||
| GenBank mRNA | BC132174.1 | - | 1 | 799 | 40130451 | |
| BC132176.1 | - | 1 | 799 | 40130453 | ||