siRNA Details
Gene Symbol: Olfr639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147084.1 | NP_667295.1 | 1 | 636 | ||
GenBank mRNA | BC145837.1 | - | 1 | 686 | 40131140 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147084.1 | NP_667295.1 | 1 | 636 | ||
GenBank mRNA | BC145837.1 | - | 1 | 686 | 40131140 |