siRNA Details
| Gene Symbol: Olfr571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104118.1 | - | 1 | 117 | 40044090 | |
| BC104119.1 | - | 1 | 117 | 40044091 | ||
| BC119420.1 | - | 1 | 124 | 40044095 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr571| Gene Symbol: Olfr571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104118.1 | - | 1 | 117 | 40044090 | |
| BC104119.1 | - | 1 | 117 | 40044091 | ||
| BC119420.1 | - | 1 | 124 | 40044095 | ||