siRNA Details
| Gene Symbol: Olfr569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147088.1 | NP_667299.1 | 1 | 126 | ||
| GenBank mRNA | BC126864.1 | - | 1 | 126 | 40042577 | |
| BC126865.1 | - | 1 | 126 | 40042578 | ||
| BC127048.1 | - | 1 | 126 | 40056696 | ||

Olfr569
Genomic Map