siRNA Details
| Gene Symbol: Olfr572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147089.2 | NP_667300.1 | 1 | 751 | ||
| GenBank mRNA | BC128002.1 | - | 1 | 755 | 40135902 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr572| Gene Symbol: Olfr572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147089.2 | NP_667300.1 | 1 | 751 | ||
| GenBank mRNA | BC128002.1 | - | 1 | 755 | 40135902 | |