siRNA Details
Gene Symbol: Olfr568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147091.2 | NP_667302.2 | 1 | 889 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147091.2 | NP_667302.2 | 1 | 889 |