siRNA Details
| Gene Symbol: Olfr561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147092.1 | NP_667303.1 | 1 | 404 | ||
| GenBank mRNA | BC120569.1 | - | 1 | 435 | 40129492 | |
| BC120571.1 | - | 1 | 435 | 40129494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr561| Gene Symbol: Olfr561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147092.1 | NP_667303.1 | 1 | 404 | ||
| GenBank mRNA | BC120569.1 | - | 1 | 435 | 40129492 | |
| BC120571.1 | - | 1 | 435 | 40129494 | ||