siRNA Details
| Gene Symbol: Olfr558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147093.3 | NP_667304.1 | 2 | 241 | ||
| GenBank mRNA | AK141545.1 | - | 2 | 241 | ||
| AY834217.1 | - | 1 | 112 | |||
| BC106820.2 | - | 1 | 107 | |||
| BC130658.1 | - | 1 | 112 | 40043445 | ||

Olfr558
Genomic Map