siRNA Details
| Gene Symbol: Olfr628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147097.2 | NP_667308.2 | 1 | 235 | ||
| GenBank mRNA | BC109148.1 | - | 1 | 321 | 40046009 | |
| BC109149.1 | - | 1 | 321 | 40046010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr628| Gene Symbol: Olfr628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147097.2 | NP_667308.2 | 1 | 235 | ||
| GenBank mRNA | BC109148.1 | - | 1 | 321 | 40046009 | |
| BC109149.1 | - | 1 | 321 | 40046010 | ||