siRNA Details
| Gene Symbol: Olfr616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147099.2 | NP_667310.2 | 1 | 428 | ||
| GenBank mRNA | BC106821.1 | - | 1 | 428 | 40043454 | |
| BC106822.1 | - | 1 | 428 | 40043455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr616| Gene Symbol: Olfr616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147099.2 | NP_667310.2 | 1 | 428 | ||
| GenBank mRNA | BC106821.1 | - | 1 | 428 | 40043454 | |
| BC106822.1 | - | 1 | 428 | 40043455 | ||