siRNA Details
| Gene Symbol: Olfr613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147100.3 | NP_667311.3 | 1 | 1497 | ||
| GenBank mRNA | AK156691.1 | - | 1 | 1496 | ||
| BC145889.1 | - | 1 | 109 | 40131192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr613| Gene Symbol: Olfr613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147100.3 | NP_667311.3 | 1 | 1497 | ||
| GenBank mRNA | AK156691.1 | - | 1 | 1496 | ||
| BC145889.1 | - | 1 | 109 | 40131192 | ||