siRNA Details
| Gene Symbol: Olfr549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147101.2 | NP_667312.2 | 1 | 664 | ||
| GenBank mRNA | BC100691.1 | - | 1 | 663 | ||
| BC100692.1 | - | 1 | 663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr549| Gene Symbol: Olfr549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147101.2 | NP_667312.2 | 1 | 664 | ||
| GenBank mRNA | BC100691.1 | - | 1 | 663 | ||
| BC100692.1 | - | 1 | 663 | |||