siRNA Details
| Gene Symbol: Olfr978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147105.2 | NP_667316.2 | 1 | 524 | ||
| GenBank mRNA | BC114537.1 | - | 1 | 523 | ||
| Gene Symbol: Olfr979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147108.2 | NP_667319.1 | 1 | 549 | ||
| GenBank mRNA | BC116754.1 | - | 1 | 549 | 40126073 | |
| BC116756.1 | - | 1 | 549 | 40126075 | ||

Olfr978
Genomic Map