siRNA Details
| Gene Symbol: Olfr978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147105.2 | NP_667316.2 | 1 | 226 | ||
| GenBank mRNA | BC114537.1 | - | 1 | 225 | ||
| Gene Symbol: Olfr980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147106.2 | NP_667317.1 | 1 | 308 | ||
| GenBank mRNA | BC116814.1 | - | 1 | 308 | 40126133 | |
| BC119187.1 | - | 1 | 308 | 8733936 | ||

Olfr978
Genomic Map