siRNA Details
| Gene Symbol: Olfr980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147106.2 | NP_667317.1 | 1 | 563 | ||
| GenBank mRNA | BC116814.1 | - | 1 | 563 | 40126133 | |
| BC119187.1 | - | 1 | 563 | 8733936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr980| Gene Symbol: Olfr980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147106.2 | NP_667317.1 | 1 | 563 | ||
| GenBank mRNA | BC116814.1 | - | 1 | 563 | 40126133 | |
| BC119187.1 | - | 1 | 563 | 8733936 | ||