siRNA Details
| Gene Symbol: Olfr979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147108.2 | NP_667319.1 | 1 | 705 | ||
| GenBank mRNA | BC116754.1 | - | 1 | 705 | 40126073 | |
| BC116756.1 | - | 1 | 705 | 40126075 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr979| Gene Symbol: Olfr979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147108.2 | NP_667319.1 | 1 | 705 | ||
| GenBank mRNA | BC116754.1 | - | 1 | 705 | 40126073 | |
| BC116756.1 | - | 1 | 705 | 40126075 | ||