siRNA Details
| Gene Symbol: Olfr570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147110.1 | NP_667321.1 | 1 | 817 | ||
| GenBank mRNA | BC116966.1 | - | 1 | 844 | 40126285 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr570| Gene Symbol: Olfr570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147110.1 | NP_667321.1 | 1 | 817 | ||
| GenBank mRNA | BC116966.1 | - | 1 | 844 | 40126285 | |