siRNA Details
| Gene Symbol: Olfr635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147118.2 | NP_667329.2 | 1 | 682 | ||
| GenBank mRNA | BC152334.1 | - | 1 | 686 | ||
| BC152355.1 | - | 1 | 686 | |||
| Gene Symbol: Olfr638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147120.1 | NP_667331.1 | 1 | 682 | ||
| GenBank mRNA | BC128011.1 | - | 1 | 681 | 40136021 | |
| BC152352.1 | - | 1 | 686 | 40136020 | ||

Olfr635
Genomic Map