siRNA Details
Gene Symbol: Olfr623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147122.2 | NP_667333.2 | 1 | 479 | ||
GenBank mRNA | BC119275.1 | - | 1 | 479 | 8734024 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147122.2 | NP_667333.2 | 1 | 479 | ||
GenBank mRNA | BC119275.1 | - | 1 | 479 | 8734024 |