siRNA Details
| Gene Symbol: Olfr503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011527.1 | NP_001011527.1 | 1 | 851 | ||
| Gene Symbol: Olfr658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147049.4 | NP_667260.1 | 2 | 1272 | ||
| GenBank mRNA | AK033992.1 | - | 2 | 1273 | ||
| BC104120.2 | - | 1 | 851 | 40044139 | ||
| BC104121.1 | - | 1 | 851 | |||

Olfr503
Genomic Map