siRNA Details
Gene Symbol: Olfr723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011530.2 | NP_001011530.2 | 1 | 255 | ||
GenBank mRNA | BC150980.1 | - | 1 | 255 | 9087893 |
Gene Symbol: Olfr724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146492.2 | NP_666703.2 | 1 | 255 | ||
GenBank mRNA | BC120628.1 | - | 1 | 255 | 40129551 | |
BC125538.1 | - | 1 | 255 | 40130053 |
