siRNA Details
| Gene Symbol: Olfr329-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147083.1 | - | 1 | 371 | 9056127 | |
| BC147084.1 | - | 1 | 371 | 9056147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr329-ps| Gene Symbol: Olfr329-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147083.1 | - | 1 | 371 | 9056127 | |
| BC147084.1 | - | 1 | 371 | 9056147 | ||