siRNA Details
| Gene Symbol: Olfr329-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147083.1 | - | 1 | 910 | 9056127 | |
| BC147084.1 | - | 1 | 910 | 9056147 | ||
| Gene Symbol: Olfr224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207695.1 | NP_997578.1 | 1 | 860 | ||

Olfr329-ps
Genomic Map