siRNA Details
| Gene Symbol: Olfr688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011533.2 | NP_001011533.2 | 1 | 427 | ||
| Gene Symbol: Olfr689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146750.1 | NP_666961.1 | 1 | 424 | ||
| GenBank mRNA | BC119379.1 | - | 1 | 448 | 8734128 | |

Olfr688
Genomic Map